Assay Details
| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159555.1 | NP_001153027.1 | 13 - 14 | 1710 | 150 | |
| NM_001159556.1 | NP_001153028.1 | 13 - 14 | 1570 | 150 | ||
| NM_001159557.1 | NP_001153029.1 | 14 - 15 | 1621 | 150 | ||
| NM_001159558.1 | NP_001153030.1 | 14 - 15 | 1737 | 150 | ||
| NM_007643.4 | NP_031669.3 | 13 - 14 | 1713 | 150 | ||
| GenBank mRNA | AK052825.1 | - | 13 - 14 | 1514 | 150 | |
| AK128908.1 | - | 14 - 15 | 1514 | 150 | ||
| AK150648.1 | - | 13 - 14 | 1534 | 150 | ||
| AK150865.1 | - | 9 - 10 | 906 | 150 | ||
| AK151355.1 | - | 13 - 14 | 1549 | 150 | ||
| AK152904.1 | - | 11 - 12 | 1220 | 150 | ||
| AK153095.1 | - | 8 - 9 | 823 | 150 | ||
| AK172162.1 | - | 14 - 15 | 1559 | 150 | ||
| BC010262.1 | - | 13 - 14 | 1522 | 3481681 | 150 | |
| HQ171908.1 | - | 4 - 5 | 284 | 150 | ||
| L23108.1 | - | 13 - 14 | 1531 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd36
Genomic Map