Assay Details
| Gene Symbol: Sgce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130188.1 | NP_001123660.1 | 8 - 9 | 1262 | 115 | |
| NM_001130190.1 | NP_001123662.1 | 7 - 8 | 1154 | 115 | ||
| NM_011360.3 | NP_035490.3 | 7 - 8 | 1154 | 115 | ||
| GenBank mRNA | AB200281.1 | - | 7 - 8 | 969 | 115 | |
| AF031919.1 | - | 7 - 8 | 1010 | 115 | ||
| AF103877.1 | - | 8 - 9 | 1068 | 115 | ||
| AK076102.1 | - | 7 - 8 | 976 | 115 | ||
| AK132210.1 | - | 7 - 8 | 1031 | 115 | ||
| BC012665.1 | - | 7 - 8 | 1060 | 4013227 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgce
Genomic Map