Assay Details
| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175006.2 | NP_778171.2 | 2 - 3 | 617 | 76 | |
| GenBank mRNA | AK083772.1 | - | 2 - 3 | 617 | 76 | |
| BC094915.1 | - | 2 - 3 | 632 | 6435959 | 76 | |
| BC138151.1 | - | 2 - 3 | 536 | 8861170 | 76 | |
| BG807013.1 | - | 2 - 3 | 267 | 76 | ||
More Information
Set Membership: |
|

Pou6f2
Genomic Map