Assay Details
| Gene Symbol: Mrpl48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003559.1 | 1 - 2 | 104 | 77 | ||
| GenBank mRNA | AK007653.1 | - | 1 - 2 | 410 | 77 | |
| BE995503.1 | - | 1 - 2 | 104 | 77 | ||
| CD363582.1 | - | 1 - 2 | 70 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB6A, member RAS oncogene family |
Rab6a |
2610028L11Rik, AA419671, Rab6 |
Anti-sense | Chr. 7 - 100607586 - 100641268 on NCBI Build GRCm38 | |
| Downstream |
pleckstrin homology domain containing, family B (evectins) member 1 |
Plekhb1 |
AI315628, PHR1, Phret1, evt-1 |
35.9kb | Sense | Chr. 7 - 100643896 - 100662394 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl48
Genomic Map