Assay Details
| Gene Symbol: Arhgap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287530.1 | NP_001274459.1 | 6 - 7 | 1095 | 64 | |
| NM_009707.4 | NP_033837.2 | 6 - 7 | 2463 | 64 | ||
| NM_178754.3 | NP_848869.1 | 6 - 7 | 1006 | 64 | ||
| NR_109847.1 | 6 - 7 | 1006 | 64 | |||
| GenBank mRNA | AF012273.2 | - | 1585 | 64 | ||
| AF177664.1 | - | 1585 | 64 | |||
| AK048162.1 | - | 6 - 7 | 960 | 64 | ||
| AK048507.1 | - | 6 - 7 | 961 | 64 | ||
| AK077996.1 | - | 6 - 7 | 968 | 64 | ||
| AK079651.1 | - | 6 - 7 | 919 | 64 | ||
| AK166015.1 | - | 6 - 7 | 926 | 64 | ||
| AK170575.1 | - | 6 - 7 | 962 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap6
Genomic Map