Assay Details
| Gene Symbol: Tpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013861.3 | NP_038889.1 | 7 - 8 | 562 | 116 | |
| GenBank mRNA | AB027568.1 | - | 7 - 8 | 563 | 116 | |
| AK028431.1 | - | 7 - 8 | 585 | 116 | ||
| AK033192.1 | - | 5 - 6 | 666 | 116 | ||
| AK034521.1 | - | 572 | 116 | |||
| AK035044.1 | - | 7 - 8 | 563 | 116 | ||
| AK052225.1 | - | 7 - 8 | 554 | 116 | ||
| AK087844.1 | - | 7 - 8 | 666 | 116 | ||
| BC015246.1 | - | 7 - 8 | 579 | 4221866 | 116 | |
More Information
Set Membership: |
|

Tpk1
Genomic Map