Assay Details
| Gene Symbol: Tpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013861.3 | NP_038889.1 | 3 - 4 | 171 | 110 | |
| GenBank mRNA | AB027568.1 | - | 3 - 4 | 172 | 110 | |
| AK028431.1 | - | 3 - 4 | 194 | 110 | ||
| AK035044.1 | - | 3 - 4 | 172 | 110 | ||
| AK052225.1 | - | 3 - 4 | 162 | 111 | ||
| AK087844.1 | - | 3 - 4 | 275 | 110 | ||
| AK136486.1 | - | 3 - 4 | 172 | 110 | ||
| BC015246.1 | - | 3 - 4 | 188 | 4221866 | 110 | |
| BC023354.1 | - | 3 - 4 | 147 | 5253545 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tpk1
Genomic Map