Assay Details
| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027164.1 | NP_081440.1 | 1 - 2 | 281 | 126 | |
| GenBank mRNA | BC050816.1 | - | 1 - 2 | 281 | 6774506 | 126 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC050816.1, NM_027164.1 transcripts.
Set Membership: |
|

Lrrc27
Genomic Map