Assay Details
| Gene Symbol: 4933432K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102286.1 | 1 - 2 | 82 | 61 | ||
| GenBank mRNA | AK005578.1 | - | 1 - 2 | 81 | 61 | |
| AK015302.1 | - | 83 | 61 | |||
| AK017025.1 | - | 81 | 61 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heparan sulfate (glucosamine) 3-O-sulfotransferase 2 | Hs3st2 |
6430516N12Rik, A830061E14Rik, AW491345 |
-87.4kb | Anti-sense | Chr. 7 - 121391610 - 121501768 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700025J12 gene | 1700025J12Rik | 74.0 | Anti-sense | Chr. 7 - 121589903 - 121591595 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2192532 | AK015302.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397304 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941231 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531343 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807823 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775732 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590509 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896174 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104131 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272146 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830193 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34840 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 705574 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61217 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667713 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303126 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191543 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477758 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356763 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432235 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927018 | AK046097.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927018 | AK052541.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927018 | AK078218.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927018 | AK144277.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927018 | AY541447.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927018 | BC038272.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927018 | NM_153800.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390533 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55550 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584109 | AK142688.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584109 | AK164738.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584109 | AK164778.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584109 | BC032178.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584109 | D10215.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584109 | NM_007785.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12225 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 162284 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15313 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 25431 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161146 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129272 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118736 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06347355 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01621424 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161146 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06347396 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01621465 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01621523 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129272 | - | - |
| Whole Mouse Genome G4122A | A_52_P8270 | AK005578.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P8270 | AK015302.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P8270 | AK017025.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136848 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129272 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06347396 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161146 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01621465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06032762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06228397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06493428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06622768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06210657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06384703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06048857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06553618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06250265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06480224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06516870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06491856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06250271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05919967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06293785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06100663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06563951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06629891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06234645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06652777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06491858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06048859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06399243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06357924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06161009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06095415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06652776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06456276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06637046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06541064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06234646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06488045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06608982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06262823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06536448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06354558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25474609 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1431489_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431536_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438356_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438356_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1431489_at | - | - |
| Mouse Genome 430 2.0 Array | 1431489_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431536_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431536_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700035055.mm.1 | NR_102286.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035055.mm.1 | TC0700004212.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700035056.mm.1 | NR_102286.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035056.mm.1 | TC0700004212.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700035054.mm.1 | NR_102286.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035054.mm.1 | TC0700004212.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431536_PM_at | - | - |
| Mouse Expression Set 430 | 1431489_PM_at | - | - |
| Mouse Expression Set 430 | 1431536_at | - | - |
| Mouse Expression Set 430 | 1431489_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431489_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431489_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431536_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431536_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 875161 | - | - |

4933432K03Rik
Genomic Map