Assay Details
| Gene Symbol: Kctd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172747.2 | NP_766335.1 | 3 - 4 | 677 | 73 | |
| GenBank mRNA | AF534881.3 | - | 3 - 4 | 663 | 73 | |
| AK075722.1 | - | 3 - 4 | 678 | 73 | ||
| BC079607.1 | - | 3 - 4 | 670 | 30359035 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kctd13
Genomic Map