Assay Details
| Gene Symbol: Bcl9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030256.2 | NP_084532.2 | 5 - 6 | 961 | 83 | |
| GenBank mRNA | AB128033.1 | - | 4 - 5 | 747 | 83 | |
| AK155681.1 | - | 6 - 7 | 920 | 83 | ||
| AY296058.1 | - | 5 - 6 | 961 | 83 | ||
| BC082304.1 | - | 4 - 5 | 1095 | 6856460 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcl9l
Genomic Map