Assay Details
| Gene Symbol: Bcl9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030256.2 | NP_084532.2 | 8 - 9 | 3613 | 79 | |
| GenBank mRNA | AB128033.1 | - | 7 - 8 | 3399 | 79 | |
| AY296058.1 | - | 8 - 9 | 3613 | 79 | ||
| BC072555.1 | - | 1 - 2 | 131 | 79 | ||
| BC082304.1 | - | 7 - 8 | 3747 | 6856460 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcl9l
Genomic Map