| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000004492.mm.1 |
NM_001081347.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004492.mm.1 |
NM_001081347.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004492.mm.1 |
NM_001252636.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004492.mm.1 |
NM_001252636.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004492.mm.1 |
NM_001252637.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004492.mm.1 |
NM_001252637.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004492.mm.1 |
NM_001252638.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004492.mm.1 |
NM_001252638.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004492.mm.1 |
TC1000000656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000004494.mm.1 |
NM_001081347.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004494.mm.1 |
NM_001081347.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004494.mm.1 |
NM_001252636.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004494.mm.1 |
NM_001252636.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004494.mm.1 |
NM_001252637.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004494.mm.1 |
NM_001252637.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004494.mm.1 |
NM_001252638.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004494.mm.1 |
NM_001252638.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004494.mm.1 |
TC1000000656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000004493.mm.1 |
NM_001081347.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004493.mm.1 |
NM_001081347.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004493.mm.1 |
NM_001252636.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004493.mm.1 |
NM_001252636.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004493.mm.1 |
NM_001252637.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004493.mm.1 |
NM_001252637.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004493.mm.1 |
NM_001252638.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004493.mm.1 |
NM_001252638.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004493.mm.1 |
TC1000000656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000004489.mm.1 |
NM_001252638.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004489.mm.1 |
NM_001252638.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000004489.mm.1 |
TC1000000656.mm.1 |
MTA TC |