Assay Details
| Gene Symbol: Rhobtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081347.1 | NP_001074816.1 | 6 - 7 | 1597 | 75 | |
| NM_001252636.1 | NP_001239565.1 | 6 - 7 | 1580 | 75 | ||
| NM_001252637.1 | NP_001239566.1 | 6 - 7 | 1701 | 75 | ||
| NM_001252638.1 | NP_001239567.1 | 7 - 8 | 1760 | 75 | ||
| GenBank mRNA | AK005770.1 | - | 6 - 7 | 1580 | 75 | |
| AK034826.1 | - | 6 - 7 | 1597 | 75 | ||
| AK078306.1 | - | 5 - 6 | 1534 | 75 | ||
| AK173015.1 | - | 1 - 2 | 961 | 75 | ||
| BC099973.1 | - | 6 - 7 | 1578 | 6432524 | 75 | |
| BC137646.1 | - | 5 - 6 | 1531 | 8860665 | 75 | |
| BC137647.1 | - | 5 - 6 | 1531 | 8860666 | 75 | |
| BC144800.1 | - | 6 - 7 | 1590 | 9053348 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhobtb1
Genomic Map