Assay Details
| Gene Symbol: Dcun1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029775.2 | NP_084051.1 | 4 - 5 | 571 | 113 | |
| GenBank mRNA | AK012764.1 | - | 4 - 5 | 583 | 113 | |
| AK013933.1 | - | 4 - 5 | 589 | 113 | ||
| AK014737.1 | - | 4 - 5 | 613 | 113 | ||
| AK134862.1 | - | 618 | 113 | |||
| BC020048.1 | - | 4 - 5 | 607 | 113 | ||
| BC020089.1 | - | 4 - 5 | 571 | 4013755 | 113 | |
| BC049641.1 | - | 760 | 113 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dcun1d5
Genomic Map