Assay Details
| Gene Symbol: Arid3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288625.1 | NP_001275554.1 | 7 - 8 | 1826 | 82 | |
| NM_001288626.1 | NP_001275555.1 | 7 - 8 | 1974 | 82 | ||
| NM_007880.4 | NP_031906.1 | 7 - 8 | 1980 | 82 | ||
| GenBank mRNA | AK141283.1 | - | 7 - 8 | 1827 | 82 | |
| AK154956.1 | - | 7 - 8 | 1952 | 82 | ||
| BC050925.1 | - | 7 - 8 | 1878 | 6831312 | 82 | |
| U60335.1 | - | 6 - 7 | 1768 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arid3a
Genomic Map