Assay Details
| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144528.3 | NP_653111.1 | 2 - 3 | 260 | 94 | |
| NR_027505.2 | 2 - 3 | 137 | 94 | |||
| GenBank mRNA | AK042319.1 | - | 2 - 3 | 127 | 94 | |
| AK154781.1 | - | 2 - 3 | 226 | 94 | ||
| BB870939.1 | - | 2 - 3 | 229 | 94 | ||
| BC016543.1 | - | 2 - 3 | 232 | 3158083 | 94 | |
| BU147682.1 | - | 2 - 3 | 324 | 94 | ||
| DV648665.1 | - | 2 - 3 | 155 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf126
Genomic Map