Assay Details
| Gene Symbol: Cldnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045517.1 | 1 - 2 | 178 | 68 | ||
| NR_045518.1 | 1 - 2 | 178 | 68 | |||
| GenBank mRNA | AK003808.1 | - | 1 - 2 | 70 | 68 | |
| AK152121.1 | - | 1 - 2 | 73 | 68 | ||
| AK152722.1 | - | 1 - 2 | 68 | 68 | ||
| BY122667.1 | - | 1 - 2 | 179 | 68 | ||
| CD560460.1 | - | 1 - 2 | 59 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003808.1, AK152121.1, AK152722.1 transcripts.
Set Membership: |
|

Cldnd1
Genomic Map