Assay Details
| Gene Symbol: Ildr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285788.1 | NP_001272717.1 | 6 - 7 | 959 | 73 | |
| NM_134109.2 | NP_598870.1 | 6 - 7 | 959 | 73 | ||
| GenBank mRNA | AK035504.1 | - | 6 - 7 | 888 | 73 | |
| AK036844.1 | - | 6 - 7 | 885 | 73 | ||
| AK136284.1 | - | 6 - 7 | 959 | 73 | ||
| BC010485.1 | - | 6 - 7 | 899 | 3483341 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ildr1
Genomic Map