Assay Details
| Gene Symbol: Atp8b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026094.3 | NP_080370.2 | 10 - 11 | 912 | 99 | |
| GenBank mRNA | AY364445.1 | - | 10 - 11 | 912 | 99 | |
| BC118977.1 | - | 10 - 11 | 912 | 40098417 | 99 | |
| BC118978.1 | - | 10 - 11 | 912 | 40098419 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp8b3
Genomic Map