Assay Details
| Gene Symbol: Kiss1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053244.5 | NP_444474.1 | 1 - 2 | 1952 | 75 | |
| GenBank mRNA | AF343726.1 | - | 1 - 2 | 249 | 75 | |
| AK039628.1 | - | 1 - 2 | 1953 | 75 | ||
| BC016531.1 | - | 1 - 2 | 395 | 3488082 | 75 | |
| EU532436.1 | - | 1 - 2 | 249 | 75 | ||
| EU532437.1 | - | 1 - 2 | 249 | 75 | ||
| EU879091.1 | - | 1 - 2 | 249 | 75 | ||
| GU075727.1 | - | 1 - 2 | 249 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kiss1r
Genomic Map