Assay Details
| Gene Symbol: Cwc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023153.3 | NP_075642.1 | 4 - 5 | 505 | 67 | |
| GenBank mRNA | AJ250394.1 | - | 4 - 5 | 396 | 67 | |
| AK002864.1 | - | 4 - 5 | 444 | 67 | ||
| AK003202.1 | - | 4 - 5 | 437 | 67 | ||
| AK003591.1 | - | 2 - 3 | 147 | 67 | ||
| AK003834.2 | - | 4 - 5 | 450 | 67 | ||
| AK145249.1 | - | 4 - 5 | 445 | 67 | ||
| AK167776.1 | - | 4 - 5 | 425 | 67 | ||
| AK168365.1 | - | 4 - 5 | 425 | 67 | ||
| AY842452.1 | - | 3 - 4 | 351 | 67 | ||
| BC004726.1 | - | 4 - 5 | 441 | 3582653 | 67 | |
| BQ840384.1 | - | 4 - 5 | 505 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cwc15
Genomic Map