Assay Details
| Gene Symbol: Slc17a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276452.1 | NP_001263381.1 | 4 - 5 | 609 | 81 | |
| NM_172773.3 | NP_766361.1 | 4 - 5 | 687 | 81 | ||
| GenBank mRNA | AK029102.1 | - | 4 - 5 | 670 | 81 | |
| AK087395.1 | - | 4 - 5 | 686 | 81 | ||
| AK169868.1 | - | 4 - 5 | 636 | 81 | ||
| BC058785.1 | - | 4 - 5 | 579 | 6309785 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc17a5
Genomic Map