Assay Details
| Gene Symbol: Leo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039522.1 | NP_001034611.1 | 9 - 10 | 1673 | 68 | |
| GenBank mRNA | AK153933.1 | - | 9 - 10 | 1648 | 68 | |
| BC082540.1 | - | 7 - 8 | 4050 | 68 | ||
| BC083358.1 | - | 9 - 10 | 1661 | 30625386 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Leo1
Genomic Map