Assay Details
| Gene Symbol: Leo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039522.1 | NP_001034611.1 | 10 - 11 | 1853 | 112 | |
| GenBank mRNA | BC082540.1 | - | 8 - 9 | 4230 | 112 | |
| BC083358.1 | - | 10 - 11 | 1841 | 30625386 | 112 | |
More Information
Set Membership: |
|

Leo1
Genomic Map