Assay Details
| Gene Symbol: Dohh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133964.2 | NP_598725.2 | 4 - 5 | 796 | 78 | |
| GenBank mRNA | AK004052.1 | - | 4 - 5 | 761 | 78 | |
| AK154104.1 | - | 4 - 5 | 796 | 78 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27289288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27289287 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345902 | BC002295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345902 | BB753904.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345902 | NM_133964.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345902 | AA000750.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345902 | AK004052.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345902 | BC002295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345902 | BB753904.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345902 | NM_133964.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345902 | AA000750.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345902 | AK004052.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P35961 | AK080664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P35961 | AK080664.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67532 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160315 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345902 | BC002295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345902 | BB753904.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345902 | NM_133964.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345902 | AA000750.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345902 | AK004052.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345902 | BC002295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345902 | BB753904.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345902 | NM_133964.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345902 | AA000750.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345902 | AK004052.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27289289 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160315 | - | - |
| Whole Mouse Genome G4122A | A_52_P35961 | AK080664.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P35961 | AK080664.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345902 | BC002295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345902 | BB753904.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345902 | NM_133964.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345902 | AA000750.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345902 | AK004052.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345902 | BC002295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345902 | BB753904.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345902 | NM_133964.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345902 | AA000750.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345902 | AK004052.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116736 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160315 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08751032 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08445329 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08751034 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08206445 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08206444 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08206448 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08751029 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27289287 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27289288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27289282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08970841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08973472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08258327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08445330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08258332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08258324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08220925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27289318 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10365122 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365119 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365121 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417322_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436402_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10365116 | - | - |
| Mouse Genome 430 2.0 Array | 1417322_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436402_at | - | - |
| Mouse Genome 430 2.0 Array | 1436402_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417322_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102115_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102115_at | - | - |
| Mouse Expression Set 430 | 1417322_PM_at | - | - |
| Mouse Expression Set 430 | 1436402_at | - | - |
| Mouse Expression Set 430 | 1417322_at | - | - |
| Mouse Expression Set 430 | 1436402_PM_at | - | - |
| Murine Genome U74v2 Set | 99078_PM_at | - | - |
| Murine Genome U74v2 Set | 99078_at | - | - |
| Murine Genome U74v2 Set | 170932_at | - | - |
| Murine Genome U74v2 Set | 170932_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417322_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436402_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417322_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436402_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436402_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436402_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417322_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417322_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 662321 | - | - |
| ABI Mouse Genome Survey Microarray | 851022 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106940152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3450184 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106940152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003450184 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2657462 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2657462 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2464955 | - | - |
| mouse-6 v1.1 (Illumina) | scl00102115.2_247-S | - | - |
| mouse-6 v1.1 (Illumina) | scl38649.8.1_81-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106940152 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3450184 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00102115.2_247-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3450184 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106940152 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2464955 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00102115.2_247-S | - | - |

Dohh
Genomic Map