Assay Details
| Gene Symbol: Arid5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023598.2 | NP_076087.2 | 8 - 9 | 1217 | 88 | |
| GenBank mRNA | AF169968.1 | - | 1448 | 88 | ||
| AF280065.1 | - | 8 - 9 | 1231 | 88 | ||
| AF401550.1 | - | 5 - 6 | 609 | 88 | ||
| AK029818.1 | - | 9 - 10 | 1349 | 88 | ||
| AK136201.1 | - | 7 - 8 | 943 | 88 | ||
| CD543091.1 | - | 1 - 2 | 91 | 88 | ||
| CN665657.1 | - | 3 - 4 | 179 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arid5b
Genomic Map