Assay Details
| Gene Symbol: Arid5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023598.2 | NP_076087.2 | 9 - 10 | 1419 | 121 | |
| GenBank mRNA | AF169968.1 | - | 1650 | 121 | ||
| AF280065.1 | - | 9 - 10 | 1433 | 121 | ||
| AF401550.1 | - | 6 - 7 | 811 | 121 | ||
| AK029818.1 | - | 10 - 11 | 1551 | 121 | ||
| AK136201.1 | - | 8 - 9 | 1145 | 121 | ||
| CD543091.1 | - | 2 - 3 | 293 | 121 | ||
| CN665657.1 | - | 4 - 5 | 381 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arid5b
Genomic Map