Assay Details
| Gene Symbol: Tjp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282095.1 | NP_001269024.1 | 18 - 19 | 2428 | 60 | |
| NM_001282096.1 | NP_001269025.1 | 18 - 19 | 2491 | 60 | ||
| NM_013769.3 | NP_038797.2 | 18 - 19 | 2530 | 60 | ||
| GenBank mRNA | AK008855.1 | - | 6 - 7 | 883 | 60 | |
| AK029083.1 | - | 18 - 19 | 2477 | 60 | ||
| AK134100.1 | - | 18 - 19 | 2531 | 60 | ||
| AK167587.1 | - | 18 - 19 | 2431 | 60 | ||
| BC012518.1 | - | 18 - 19 | 2495 | 3600848 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tjp3
Genomic Map