Assay Details
| Gene Symbol: Tjp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282095.1 | NP_001269024.1 | 19 - 20 | 2687 | 66 | |
| NM_001282096.1 | NP_001269025.1 | 19 - 20 | 2750 | 66 | ||
| NM_013769.3 | NP_038797.2 | 19 - 20 | 2789 | 66 | ||
| GenBank mRNA | AF157006.1 | - | 19 - 20 | 2760 | 66 | |
| AK008855.1 | - | 7 - 8 | 1142 | 66 | ||
| AK029083.1 | - | 19 - 20 | 2736 | 66 | ||
| AK134100.1 | - | 19 - 20 | 2790 | 66 | ||
| BC012518.1 | - | 19 - 20 | 2754 | 3600848 | 66 | |
| BG228652.1 | - | 2 - 3 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tjp3
Genomic Map