Assay Details
| Gene Symbol: Sgta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024499.1 | NP_078775.1 | 4 - 5 | 360 | 61 | |
| GenBank mRNA | AK039966.1 | - | 3 - 4 | 355 | 61 | |
| AK051005.1 | - | 4 - 5 | 359 | 61 | ||
| AK079168.1 | - | 4 - 5 | 370 | 61 | ||
| AK165557.1 | - | 4 - 5 | 384 | 61 | ||
| BC003836.1 | - | 4 - 5 | 360 | 3486148 | 61 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK003018.1 | - | Chr. - - [-] on Build | 61 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sgta
Genomic Map