Assay Details
| Gene Symbol: Sgta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024499.1 | NP_078775.1 | 7 - 8 | 697 | 74 | |
| GenBank mRNA | AK039966.1 | - | 6 - 7 | 692 | 74 | |
| AK051005.1 | - | 7 - 8 | 696 | 74 | ||
| AK165557.1 | - | 7 - 8 | 721 | 74 | ||
| BC003836.1 | - | 7 - 8 | 697 | 3486148 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgta
Genomic Map