Assay Details
| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163780.1 | NP_001157252.1 | 10 - 11 | 1799 | 91 | |
| NM_019987.2 | NP_064371.2 | 10 - 11 | 1727 | 91 | ||
| GenBank mRNA | AF225918.1 | - | 10 - 11 | 1567 | 91 | |
| AK052178.1 | - | 3 - 4 | 696 | 91 | ||
| AK087484.1 | - | 10 - 11 | 1651 | 91 | ||
| AK133537.1 | - | 10 - 11 | 1666 | 91 | ||
| AK154185.1 | - | 10 - 11 | 1800 | 91 | ||
| AK173068.1 | - | 5 - 6 | 956 | 91 | ||
| BC028863.1 | - | 9 - 10 | 1484 | 4224269 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ick
Genomic Map