Assay Details
| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163780.1 | NP_001157252.1 | 11 - 12 | 1952 | 64 | |
| NM_019987.2 | NP_064371.2 | 11 - 12 | 1880 | 64 | ||
| GenBank mRNA | AF225918.1 | - | 11 - 12 | 1720 | 64 | |
| AK052178.1 | - | 4 - 5 | 849 | 64 | ||
| AK087484.1 | - | 11 - 12 | 1804 | 64 | ||
| AK133537.1 | - | 11 - 12 | 1819 | 64 | ||
| AK154185.1 | - | 11 - 12 | 1953 | 64 | ||
| AK173068.1 | - | 6 - 7 | 1109 | 64 | ||
| BC028863.1 | - | 10 - 11 | 1637 | 4224269 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ick
Genomic Map