Assay Details
| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163780.1 | NP_001157252.1 | 12 - 13 | 2081 | 96 | |
| NM_019987.2 | NP_064371.2 | 12 - 13 | 2009 | 96 | ||
| GenBank mRNA | AF225918.1 | - | 12 - 13 | 1849 | 96 | |
| AK052178.1 | - | 5 - 6 | 978 | 96 | ||
| AK087484.1 | - | 12 - 13 | 1933 | 96 | ||
| AK154185.1 | - | 12 - 13 | 2082 | 96 | ||
| AK173068.1 | - | 7 - 8 | 1238 | 96 | ||
| AK197692.1 | - | 1 - 2 | 59 | 96 | ||
| BC028863.1 | - | 11 - 12 | 1766 | 4224269 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ick
Genomic Map