Assay Details
| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163780.1 | NP_001157252.1 | 3 - 4 | 620 | 102 | |
| NM_019987.2 | NP_064371.2 | 3 - 4 | 548 | 102 | ||
| GenBank mRNA | AF225918.1 | - | 3 - 4 | 388 | 102 | |
| AK087484.1 | - | 3 - 4 | 471 | 102 | ||
| AK133537.1 | - | 3 - 4 | 487 | 102 | ||
| AK154185.1 | - | 3 - 4 | 621 | 102 | ||
| AK206365.1 | - | 2 - 3 | 201 | 102 | ||
| BC028863.1 | - | 2 - 3 | 305 | 4224269 | 102 | |
More Information
Set Membership: |
|

Ick
Genomic Map