Assay Details
| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163780.1 | NP_001157252.1 | 5 - 6 | 826 | 68 | |
| NM_019987.2 | NP_064371.2 | 5 - 6 | 754 | 68 | ||
| GenBank mRNA | AF225918.1 | - | 5 - 6 | 594 | 68 | |
| AK087484.1 | - | 5 - 6 | 677 | 68 | ||
| AK133537.1 | - | 5 - 6 | 693 | 68 | ||
| AK154185.1 | - | 5 - 6 | 827 | 68 | ||
| BC028863.1 | - | 4 - 5 | 511 | 4224269 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ick
Genomic Map