Assay Details
| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163780.1 | NP_001157252.1 | 7 - 8 | 1133 | 110 | |
| NM_019987.2 | NP_064371.2 | 7 - 8 | 1061 | 110 | ||
| GenBank mRNA | AF225918.1 | - | 7 - 8 | 901 | 110 | |
| AK087484.1 | - | 7 - 8 | 984 | 110 | ||
| AK133537.1 | - | 7 - 8 | 1000 | 110 | ||
| AK154185.1 | - | 7 - 8 | 1134 | 110 | ||
| AK173068.1 | - | 2 - 3 | 290 | 110 | ||
| BC028863.1 | - | 6 - 7 | 818 | 4224269 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ick
Genomic Map