Assay Details
| Gene Symbol: Gclc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010295.2 | NP_034425.1 | 13 - 14 | 1756 | 97 | |
| GenBank mRNA | AK144339.1 | - | 13 - 14 | 1757 | 97 | |
| AK160876.1 | - | 13 - 14 | 1516 | 97 | ||
| AK169676.1 | - | 13 - 14 | 1729 | 97 | ||
| BC019374.1 | - | 13 - 14 | 1513 | 4195425 | 97 | |
| U85414.1 | - | 13 - 14 | 1481 | 97 | ||
| U85498.1 | - | 13 - 14 | 1481 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gclc
Genomic Map