| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700004958.mm.1 |
NM_019546.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004958.mm.1 |
NM_019546.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004958.mm.1 |
TC0700000564.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004959.mm.1 |
NM_019546.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004959.mm.1 |
NM_019546.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004959.mm.1 |
TC0700000564.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004957.mm.1 |
NM_019546.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004957.mm.1 |
NM_019546.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004957.mm.1 |
TC0700000564.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004960.mm.1 |
NM_019546.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004960.mm.1 |
NM_019546.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004960.mm.1 |
TC0700000564.mm.1 |
MTA TC |