Assay Details
| Gene Symbol: Pfkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008826.4 | NP_032852.2 | 21 - 22 | 2310 | 80 | |
| GenBank mRNA | AK030511.1 | - | 21 - 22 | 2265 | 80 | |
| AK030892.1 | - | 14 - 15 | 1409 | 80 | ||
| AK036318.1 | - | 21 - 22 | 2243 | 80 | ||
| AK081313.1 | - | 21 - 22 | 2261 | 80 | ||
| AK159328.1 | - | 21 - 22 | 2255 | 80 | ||
| AK168660.1 | - | 21 - 22 | 2266 | 80 | ||
| BC020097.1 | - | 21 - 22 | 2264 | 4014395 | 80 | |
| BC029336.1 | - | 21 - 22 | 2253 | 80 | ||
| J03928.1 | - | 21 - 22 | 2239 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pfkl
Genomic Map