Assay Details
| Gene Symbol: Rrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010925.2 | NP_035055.2 | 11 - 12 | 1274 | 95 | |
| GenBank mRNA | AF312394.1 | - | 10 - 11 | 1037 | 95 | |
| AK077540.1 | - | 11 - 12 | 1199 | 95 | ||
| BC003913.1 | - | 4 - 5 | 370 | 95 | ||
| BC022910.1 | - | 10 - 11 | 1140 | 4165783 | 95 | |
| BC065993.1 | - | 11 - 12 | 1236 | 5684672 | 95 | |
| BC065994.1 | - | 11 - 12 | 1194 | 5685269 | 95 | |
| U79773.1 | - | 12 - 13 | 1073 | 95 | ||
| U79774.1 | - | 11 - 12 | 1226 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC003913.1 transcripts.
Set Membership: |
|

Rrp1
Genomic Map