Assay Details
| Gene Symbol: Rrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010925.2 | NP_035055.2 | 7 - 8 | 714 | 82 | |
| GenBank mRNA | AF312394.1 | - | 6 - 7 | 477 | 82 | |
| AK077540.1 | - | 7 - 8 | 639 | 82 | ||
| BC022910.1 | - | 6 - 7 | 580 | 4165783 | 82 | |
| BC065993.1 | - | 7 - 8 | 676 | 5684672 | 82 | |
| BC065994.1 | - | 7 - 8 | 634 | 5685269 | 82 | |
| U79773.1 | - | 7 - 8 | 666 | 82 | ||
| U79774.1 | - | 7 - 8 | 666 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rrp1
Genomic Map