Assay Details
| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CB589155.1 | - | 62 | 60 | ||
| CX201977.1 | - | 1 - 2 | 94 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dedd2| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CB589155.1 | - | 62 | 60 | ||
| CX201977.1 | - | 1 - 2 | 94 | 60 | ||
Set Membership: |
|