Assay Details
| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207677.3 | NP_997560.3 | 3 - 4 | 589 | 101 | |
| GenBank mRNA | AF457576.1 | - | 2 - 3 | 454 | 101 | |
| AF543541.1 | - | 2 - 3 | 454 | 101 | ||
| AK011713.1 | - | 2 - 3 | 295 | 101 | ||
| BC037043.1 | - | 1 - 2 | 133 | 101 | ||
| BC092250.1 | - | 3 - 4 | 589 | 5043318 | 101 | |
| CB589155.1 | - | 554 | 101 | |||
| CX201206.1 | - | 2 - 3 | 441 | 101 | ||
More Information
Set Membership: |
|

Dedd2
Genomic Map