Assay Details
| Gene Symbol: Sgpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009163.3 | NP_033189.2 | 2 - 3 | 222 | 136 | |
| GenBank mRNA | AF036894.1 | - | 1 - 2 | 92 | 136 | |
| AK036747.1 | - | 2 - 3 | 110 | 136 | ||
| AK037789.1 | - | 2 - 3 | 107 | 136 | ||
| AK043024.1 | - | 2 - 3 | 109 | 136 | ||
| AK049342.1 | - | 2 - 3 | 130 | 136 | ||
| AK150053.1 | - | 2 - 3 | 117 | 136 | ||
| AK173137.1 | - | 2 - 3 | 151 | 136 | ||
| BC026135.1 | - | 2 - 3 | 168 | 4212293 | 136 | |
| BY061956.1 | - | 2 - 3 | 208 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF036894.1, AK036747.1, AK037789.1, AK043024.1, AK049342.1, AK150053.1, BC026135.1, NM_009163.3 transcripts.
Set Membership: |
|

Sgpl1
Genomic Map