Assay Details
| Gene Symbol: Dapk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010019.3 | NP_034149.2 | 7 - 8 | 783 | 71 | |
| GenBank mRNA | AB018002.1 | - | 7 - 8 | 773 | 71 | |
| AF052942.1 | - | 5 - 6 | 463 | 71 | ||
| BX527735.1 | - | 3 - 4 | 167 | 71 | ||
| GU109477.1 | - | 7 - 8 | 783 | 71 | ||
More Information
Set Membership: |
|

Dapk2
Genomic Map