Assay Details
| Gene Symbol: Pdgfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027924.2 | NP_082200.1 | 3 - 4 | 694 | 75 | |
| GenBank mRNA | AF335583.1 | - | 3 - 4 | 511 | 75 | |
| AK003359.2 | - | 3 - 4 | 696 | 75 | ||
| AK141551.1 | - | 3 - 4 | 705 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfd
Genomic Map