Assay Details
| Gene Symbol: Zfp266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082485.1 | NP_001075954.1 | 4 - 5 | 499 | 87 | |
| NM_001135019.1 | NP_001128491.1 | 4 - 5 | 488 | 87 | ||
| GenBank mRNA | AK019992.1 | - | 4 - 5 | 484 | 87 | |
| BC048701.1 | - | 4 - 5 | 439 | 87 | ||
| BC061209.1 | - | 4 - 5 | 485 | 87 | ||
| BC089552.1 | - | 4 - 5 | 498 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061209.1, NM_001082485.1, NM_001135019.1 transcripts.
Set Membership: |
|

Zfp266
Genomic Map