Assay Details
| Gene Symbol: Zfp266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082485.1 | NP_001075954.1 | 5 - 6 | 584 | 65 | |
| NM_001135019.1 | NP_001128491.1 | 5 - 6 | 573 | 65 | ||
| GenBank mRNA | BC048701.1 | - | 5 - 6 | 524 | 65 | |
| BC061209.1 | - | 5 - 6 | 570 | 65 | ||
| BC089552.1 | - | 5 - 6 | 583 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp266
Genomic Map